4-[(4-Benzyl-2-bromophenoxy)methyl]piperidine hydrochloride

Supplier: Matrix Scientific

050350-500MG 050350-2.5G
101891-300EA 364.62 USD
101891-300 101891-302
4-[(4-Benzyl-2-bromophenoxy)methyl]piperidine hydrochloride
4-[(4-Benzyl-2-bromophenoxy)methyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₉H₂₃BrClNO
MDL Number: MFCD13560751
CAS Number: 1220017-58-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR