3-(2-Bromo-4-methoxyphenoxy)pyrrolidine hydrochloride

Supplier: Matrix Scientific

050341-500MG 050341-2.5G
101891-264EA 364.62 USD
101891-264 101891-266
3-(2-Bromo-4-methoxyphenoxy)pyrrolidine hydrochloride
3-(2-Bromo-4-methoxyphenoxy)pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₅BrClNO₂
MDL Number: MFCD13560742
CAS Number: 1220017-76-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR