Methyl-3-(3-pyrrolidinylmethoxy)propanoate hydrochloride

Supplier: Matrix Scientific

050336-2.5G 050336-500MG
101891-246EA 838.46 USD
101891-246 101891-244
Methyl-3-(3-pyrrolidinylmethoxy)propanoate hydrochloride
Methyl-3-(3-pyrrolidinylmethoxy)propanoate hydrochloride

Hazard: Irritant

Formula: C₉H₁₈ClNO₃
MDL Number: MFCD13560737
CAS Number: 1220017-08-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR