Methyl-3-[2-(4-piperidinyl)ethoxy]propanoate hydrochloride
Supplier: Matrix Scientific
050333-500MG
050333-2.5G
101891-234EA
838.46
USD
101891-234
101891-232
Methyl-3-[2-(4-piperidinyl)ethoxy]propanoate hydrochloride
Methyl-3-[2-(4-piperidinyl)ethoxy]propanoate hydrochloride
Hazard: Irritant
Formula:
C₁₁H₂₂ClNO₃ |
MDL Number:
MFCD13560734 CAS Number: 1220032-60-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...