3-Pyrrolidinylmethyl-5-(trifluoromethyl)-2-pyridinyl ether hydrochloride

Supplier: Matrix Scientific

050316-500MG 050316-2.5G
101891-164EA 364.62 USD
101891-164 101891-166
3-Pyrrolidinylmethyl-5-(trifluoromethyl)-2-pyridinyl ether hydrochloride
3-Pyrrolidinylmethyl-5-(trifluoromethyl)-2-pyridinyl ether hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₄ClF₃N₂O
MDL Number: MFCD13560717
CAS Number: 1220016-81-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR