Methyl-4-[(3-pyrrolidinylmethoxy)methyl]benzoate hydrochloride
Supplier: Matrix Scientific
050260-2.5G
050260-500MG
101890-942EA
838.46
USD
101890-942
101890-940
Methyl-4-[(3-pyrrolidinylmethoxy)methyl]benzoate hydrochloride
Methyl-4-[(3-pyrrolidinylmethoxy)methyl]benzoate hydrochloride
Hazard: Irritant
Formula:
C₁₄H₂₀ClNO₃ |
MDL Number:
MFCD13560661 CAS Number: 1219972-28-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...