4-[2-(2-Benzyl-4-chlorophenoxy)ethyl]piperidine hydrochloride
Supplier: Matrix Scientific
050241-2.5G
050241-500MG
101890-864EA
364.62
USD
101890-864
101890-866
4-[2-(2-Benzyl-4-chlorophenoxy)ethyl]piperidine hydrochloride
4-[2-(2-Benzyl-4-chlorophenoxy)ethyl]piperidine hydrochloride
Hazard: Irritant
Formula:
C₂₀H₂₅Cl₂NO |
MDL Number:
MFCD13560642 CAS Number: 1219972-23-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...