Benzyl-4-[2-(2-piperidinyl)ethoxy]benzoate hydrochloride
Supplier: Matrix Scientific
050235-2.5G
050235-500MG
101890-842EA
959.02
USD
101890-842
101890-840
Benzyl-4-[2-(2-piperidinyl)ethoxy]benzoate hydrochloride
Benzyl-4-[2-(2-piperidinyl)ethoxy]benzoate hydrochloride
Hazard: Irritant
Formula:
C₂₁H₂₆ClNO₃ MW: 375.9 g/mol |
MDL Number:
MFCD13560636 CAS Number: 1220030-11-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...