3-[2-(2,4-Difluorophenoxy)ethyl]piperidine hydrochloride
Supplier: Matrix Scientific
050202-500MG
050202-2.5G
101890-710EA
838.46
USD
101890-710
101890-708
3-[2-(2,4-Difluorophenoxy)ethyl]piperidine hydrochloride
3-[2-(2,4-Difluorophenoxy)ethyl]piperidine hydrochloride
Hazard: Irritant
Formula:
C₁₃H₁₈ClF₂NO |
MDL Number:
MFCD13560603 CAS Number: 1220032-30-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...