2-[2-(4-Methyl-2-nitrophenoxy)ethyl]piperidine hydrochloride
Supplier: Matrix Scientific
050187-2.5G
050187-500MG
101890-650EA
838.46
USD
101890-650
101890-648
2-[2-(4-Methyl-2-nitrophenoxy)ethyl]piperidine hydrochloride
2-[2-(4-Methyl-2-nitrophenoxy)ethyl]piperidine hydrochloride
Hazard: Irritant
Formula:
C₁₄H₂₁ClN₂O₃ |
MDL Number:
MFCD13560588 CAS Number: 1219982-41-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...