3-[2-(4-Piperidinyl)ethoxy]pyridine hydrochloride

Supplier: Matrix Scientific

050177-2.5G 050177-500MG
101890-610EA 838.46 USD
101890-610 101890-608
3-[2-(4-Piperidinyl)ethoxy]pyridine hydrochloride
3-[2-(4-Piperidinyl)ethoxy]pyridine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₉ClN₂O
MDL Number: MFCD13560578
CAS Number: 1219982-51-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR