Methyl-4-(3-pyrrolidinylmethoxy)benzoate hydrochloride

Supplier: Matrix Scientific

050131-500MG 050131-2.5G
101890-424EA 364.62 USD
101890-424 101890-426
Methyl-4-(3-pyrrolidinylmethoxy)benzoate hydrochloride
Methyl-4-(3-pyrrolidinylmethoxy)benzoate hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₈ClNO₃
MDL Number: MFCD13560533
CAS Number: 1220032-13-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR