4-[2-(3-Piperidinyl)ethoxy]phenyl propyl ether hydrochloride

Supplier: Matrix Scientific

050121-500MG
101890-384EA 272.55 USD
101890-384
4-[2-(3-Piperidinyl)ethoxy]phenyl propyl ether hydrochloride
4-[2-(3-Piperidinyl)ethoxy]phenyl propyl ether hydrochloride

Hazard: Irritant

Formula: C₁₆H₂₆ClNO₂
MDL Number: MFCD13560523
CAS Number: 1220037-60-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR