3-[2-(4-Chloro-3,5-dimethylphenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

050101-500MG 050101-2.5G
101890-304EA 364.62 USD
101890-304 101890-306
3-[2-(4-Chloro-3,5-dimethylphenoxy)ethyl]piperidine hydrochloride
3-[2-(4-Chloro-3,5-dimethylphenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₃Cl₂NO
MDL Number: MFCD13560503
CAS Number: 1219981-36-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR