3-[2-(2-Allylphenoxy)ethyl]piperidine hydrochloride
Supplier: Matrix Scientific
050021-2.5G
050021-500MG
101889-986EA
838.46
USD
101889-986
101889-984
3-[2-(2-Allylphenoxy)ethyl]piperidine hydrochloride
3-[2-(2-Allylphenoxy)ethyl]piperidine hydrochloride
Hazard: Irritant
Formula:
C₁₆H₂₄ClNO |
MDL Number:
MFCD13560423 CAS Number: 1220029-01-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...