3-[2-(2-Ethylphenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

050017-500MG 050017-2.5G
101889-968EA 364.62 USD
101889-968 101889-970
3-[2-(2-Ethylphenoxy)ethyl]piperidine hydrochloride
3-[2-(2-Ethylphenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₄ClNO
MDL Number: MFCD13560419
CAS Number: 1220037-12-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR