2-(2-Piperidinyl)ethyl-2,3,5-trimethylphenylether hydrochloride

Supplier: Matrix Scientific

050002-500MG 050002-2.5G
101889-910EA 838.46 USD
101889-910 101889-908
2-(2-Piperidinyl)ethyl-2,3,5-trimethylphenylether hydrochloride
2-(2-Piperidinyl)ethyl-2,3,5-trimethylphenylether hydrochloride

Hazard: Irritant

Formula: C₁₆H₂₆ClNO
MDL Number: MFCD13560404
CAS Number: 1220019-82-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR