3-[2-(2-Ethoxyphenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

049969-500MG 049969-2.5G
101889-776EA 272.55 USD
101889-776 101889-778
3-[2-(2-Ethoxyphenoxy)ethyl]piperidine hydrochloride
3-[2-(2-Ethoxyphenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₄ClNO₂
MDL Number: MFCD13560371
CAS Number: 1219981-50-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR