3-[2-(2-Chloro-5-methylphenoxy)ethyl]piperidine hydrochloride
Supplier: Matrix Scientific
049961-500MG
049961-2.5G
101889-744EA
272.55
USD
101889-744
101889-746
3-[2-(2-Chloro-5-methylphenoxy)ethyl]piperidine hydrochloride
3-[2-(2-Chloro-5-methylphenoxy)ethyl]piperidine hydrochloride
Hazard: Irritant
Formula:
C₁₄H₂₁Cl₂NO |
MDL Number:
MFCD13560363 CAS Number: 1219960-60-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...