4-[2-(2-Propoxyethoxy)ethyl]piperidine hydrochloride
Supplier: Matrix Scientific
049912-2.5G
049912-500MG
101889-548EA
364.62
USD
101889-548
101889-550
4-[2-(2-Propoxyethoxy)ethyl]piperidine hydrochloride
4-[2-(2-Propoxyethoxy)ethyl]piperidine hydrochloride
Hazard: Irritant
Formula:
C₁₂H₂₆ClNO₂ |
MDL Number:
MFCD13560314 CAS Number: 1220028-36-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...