4-Isopropylbenzyl-2-(3-piperidinyl)ethyl ether hydrochloride

Supplier: Matrix Scientific

049893-500MG 049893-2.5G
101889-472EA 272.55 USD
101889-472 101889-474
4-Isopropylbenzyl-2-(3-piperidinyl)ethyl ether hydrochloride
4-Isopropylbenzyl-2-(3-piperidinyl)ethyl ether hydrochloride

Hazard: Irritant

Formula: C₁₇H₂₈ClNO
MDL Number: MFCD13560295
CAS Number: 1220017-98-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR