3-[([1,1'-Biphenyl]-4-ylmethoxy)methyl]pyrrolidine hydrochloride

Supplier: Matrix Scientific

049887-500MG 049887-2.5G
101889-448EA 364.62 USD
101889-448 101889-450
3-[([1,1'-Biphenyl]-4-ylmethoxy)methyl]pyrrolidine hydrochloride
3-[([1,1'-Biphenyl]-4-ylmethoxy)methyl]pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₈H₂₂ClNO
MDL Number: MFCD13560289
CAS Number: 1219968-11-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR