4-[2-([1,1'-Biphenyl]-4-ylmethoxy)ethyl]piperidine hydrochloride
Supplier: Matrix Scientific
049884-500MG
049884-2.5G
101889-436EA
272.55
USD
101889-436
101889-438
4-[2-([1,1'-Biphenyl]-4-ylmethoxy)ethyl]piperidine hydrochloride
4-[2-([1,1'-Biphenyl]-4-ylmethoxy)ethyl]piperidine hydrochloride
Hazard: Irritant
Formula:
C₂₀H₂₆ClNO |
MDL Number:
MFCD13560286 CAS Number: 1219949-12-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...