3-[2-(Pentyloxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

049841-2.5G 049841-500MG
101889-264EA 364.62 USD
101889-264 101889-266
3-[2-(Pentyloxy)ethyl]piperidine hydrochloride
3-[2-(Pentyloxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₂H₂₆ClNO
MDL Number: MFCD13560243
CAS Number: 1219949-08-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR