3-[(2-Methoxyethoxy)methyl]pyrrolidine hydrochloride

Supplier: Matrix Scientific

049811-500MG 049811-2.5G
101889-144EA 364.62 USD
101889-144 101889-146
3-[(2-Methoxyethoxy)methyl]pyrrolidine hydrochloride
3-[(2-Methoxyethoxy)methyl]pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₈H₁₈ClNO₂
MDL Number: MFCD13560213
CAS Number: 1219967-70-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR