3-[(2-Cyclohexylethoxy)methyl]pyrrolidine hydrochloride
Supplier: Matrix Scientific
049799-2.5G
049799-500MG
101889-096EA
364.62
USD
101889-096
101889-098
3-[(2-Cyclohexylethoxy)methyl]pyrrolidine hydrochloride
3-[(2-Cyclohexylethoxy)methyl]pyrrolidine hydrochloride
Hazard: Irritant
Formula:
C₁₃H₂₆ClNO |
MDL Number:
MFCD13560201 CAS Number: 1220028-46-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...