[(2-Butoxy-1-naphthyl)methyl]amine hydrochloride

Supplier: Matrix Scientific

048781-500MG
101885-142EA 174.8 USD
101885-142
[(2-Butoxy-1-naphthyl)methyl]amine hydrochloride
[(2-Butoxy-1-naphthyl)methyl]amine hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₀ClNO
MDL Number: MFCD12195862
CAS Number: 1201633-62-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR