1-[1-(2-Furylmethyl)-3-piperidinyl]methanamine

Supplier: Matrix Scientific

048780-500MG
101885-140EA 269.23 USD
101885-140
1-[1-(2-Furylmethyl)-3-piperidinyl]methanamine
1-[1-(2-Furylmethyl)-3-piperidinyl]methanamine

Hazard: Irritant

Formula: C₁₁H₁₈N₂O
MDL Number: MFCD12195859
CAS Number: 725212-65-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR