4,4-Dimethylpentanoic acid

Supplier: Matrix Scientific

048776-500MG
101885-132EA 330.77 USD
101885-132
4,4-Dimethylpentanoic acid
4,4-Dimethylpentanoic acid

Hazard: Irritant

Formula: C₇H₁₄O₂
MDL Number: MFCD00101613
CAS Number: 1118-47-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR