[2-(3-Propoxyphenyl)ethyl]amine hydrochloride

Supplier: Matrix Scientific

048769-500MG
101885-118EA 201.25 USD
101885-118
[2-(3-Propoxyphenyl)ethyl]amine hydrochloride
[2-(3-Propoxyphenyl)ethyl]amine hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₈ClNO
MDL Number: MFCD11226489
CAS Number: 1135289-15-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR