(4-Butoxy-3-methoxybenzyl)amine

Supplier: Matrix Scientific

048763-500MG
101885-106EA 201.25 USD
101885-106
(4-Butoxy-3-methoxybenzyl)amine
(4-Butoxy-3-methoxybenzyl)amine

Hazard: Irritant

Formula: C₁₂H₁₉NO₂
MDL Number: MFCD07754790
CAS Number: 854180-43-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR