(3-Methoxy-4-propoxybenzyl)amine

Supplier: Matrix Scientific

048762-500MG
101885-104EA 201.25 USD
101885-104
(3-Methoxy-4-propoxybenzyl)amine
(3-Methoxy-4-propoxybenzyl)amine

Hazard: Irritant

Formula: C₁₁H₁₇NO₂
MDL Number: MFCD09704266
CAS Number: 854207-58-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR