(3-Methyl-1H-pyrazol-5-yl)methanol

Supplier: Matrix Scientific

Synonyms: (3-methyl-1H-pyrazol-5-yl)methanol

048753-500MG
101885-084EA 306.15 USD
101885-084
(3-Methyl-1H-pyrazol-5-yl)methanol
(3-Methyl-1H-pyrazol-5-yl)methanol

Hazard: Irritant

Formula: C₅H₈N₂O
MW: 112.13 g/mol
MDL Number: MFCD00159583
CAS Number: 17607-71-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR