2-Chlorobenzamidoxime

Supplier: Matrix Scientific

048748-500MG
101885-074EA 269.23 USD
101885-074
2-Chlorobenzamidoxime
2-Chlorobenzamidoxime

Hazard: Irritant

Formula: C₇H₇ClN₂O
MDL Number: MFCD00859262
CAS Number: 29568-74-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR