1-[1-(Cyclopropylmethyl)-4-piperidinyl]methanamine

Supplier: Matrix Scientific

048746-500MG
101885-070EA 263.08 USD
101885-070
1-[1-(Cyclopropylmethyl)-4-piperidinyl]methanamine
1-[1-(Cyclopropylmethyl)-4-piperidinyl]methanamine

Hazard: Irritant

Formula: C₁₀H₂₀N₂
MDL Number: MFCD10003421
CAS Number: 1017400-92-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR