[4-(2-Aminoethyl)phenyl]dimethylamine dihydrochloride

Supplier: Matrix Scientific

048744-500MG
101885-066EA 306.15 USD
101885-066
[4-(2-Aminoethyl)phenyl]dimethylamine dihydrochloride
[4-(2-Aminoethyl)phenyl]dimethylamine dihydrochloride

Hazard: Irritant

Formula: C₁₀H₁₈Cl₂N₂
MDL Number: MFCD11226483
CAS Number: 102880-23-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR