[1-(2-Ethoxyphenyl)ethyl]amine hydrochloride

Supplier: Matrix Scientific

048737-500MG
101885-054EA 269.23 USD
101885-054
[1-(2-Ethoxyphenyl)ethyl]amine hydrochloride
[1-(2-Ethoxyphenyl)ethyl]amine hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₆ClNO
MDL Number: MFCD10703565
CAS Number: 1135292-74-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR