6-(Dimethoxymethyl)-3-methyl-3H-imidazo[4,5-b]pyridine

Supplier: Matrix Scientific

048476-250MG
101884-034EA 393.3 USD
101884-034
6-(Dimethoxymethyl)-3-methyl-3H-imidazo[4,5-b]pyridine
6-(Dimethoxymethyl)-3-methyl-3H-imidazo[4,5-b]pyridine

Hazard: Irritant

Formula: C₁₀H₁₃N₃O₂
MDL Number: MFCD12922820
CAS Number: 1186311-14-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR