2-Bromo-6-methoxyimidazo[1,2-a]pyridine

Supplier: Matrix Scientific

048475-250MG
101884-032EA 393.3 USD
101884-032
2-Bromo-6-methoxyimidazo[1,2-a]pyridine
2-Bromo-6-methoxyimidazo[1,2-a]pyridine

Hazard: Irritant

Formula: C₈H₇BrN₂O
MDL Number: MFCD12922819
CAS Number: 1042141-33-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR