(E)-3-Methyl-3H-imidazo[4,5-b]pyridine-6-carbaldehyde oxime

Supplier: Matrix Scientific

048472-250MG
101884-026EA 393.3 USD
101884-026
(E)-3-Methyl-3H-imidazo[4,5-b]pyridine-6-carbaldehyde oxime
(E)-3-Methyl-3H-imidazo[4,5-b]pyridine-6-carbaldehyde oxime

Hazard: Irritant

Formula: C₈H₈N₄O
MDL Number: MFCD12922816
CAS Number: 1186405-20-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR