(3-Methyl-3H-imidazo[4,5-b]pyridin-6-yl)methanol

Supplier: Matrix Scientific

048471-250MG
101884-024EA 393.3 USD
101884-024
(3-Methyl-3H-imidazo[4,5-b]pyridin-6-yl)methanol
(3-Methyl-3H-imidazo[4,5-b]pyridin-6-yl)methanol

Hazard: Irritant

Formula: C₈H₉N₃O
MDL Number: MFCD12922815
CAS Number: 1186310-79-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR