3-Methyl-3H-imidazo[4,5-b]pyridine

Supplier: Matrix Scientific

048470-250MG
101884-022EA 170.2 USD
101884-022
3-Methyl-3H-imidazo[4,5-b]pyridine
3-Methyl-3H-imidazo[4,5-b]pyridine

Hazard: Irritant

Formula: C₇H₇N₃
MDL Number: MFCD03676333
CAS Number: 6688-61-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR