6-Chloro-3-methyl-3H-imidazo[4,5-b]pyridine

Supplier: Matrix Scientific

048465-250MG
101884-012EA 526.15 USD
101884-012
6-Chloro-3-methyl-3H-imidazo[4,5-b]pyridine
6-Chloro-3-methyl-3H-imidazo[4,5-b]pyridine

Hazard: Irritant

Formula: C₇H₆ClN₃
MDL Number: MFCD08692351
CAS Number: 108847-89-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR