6-Bromo-2-(chloromethyl)furo[3,2-b]pyridine

Supplier: Matrix Scientific

048457-250MG
101883-996EA 393.3 USD
101883-996
6-Bromo-2-(chloromethyl)furo[3,2-b]pyridine
6-Bromo-2-(chloromethyl)furo[3,2-b]pyridine

Hazard: Irritant

Formula: C₈H₅BrClNO
MDL Number: MFCD12922808
CAS Number: 1186310-94-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR