6-Chloro-2-(trimethylsilyl)furo[3,2-b]pyridine

Supplier: Matrix Scientific

048456-250MG
101883-994EA 393.3 USD
101883-994
6-Chloro-2-(trimethylsilyl)furo[3,2-b]pyridine
6-Chloro-2-(trimethylsilyl)furo[3,2-b]pyridine

Hazard: Irritant

Formula: C₁₀H₁₂ClNOSi
MDL Number: MFCD12922807
CAS Number: 1186310-77-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR