(trans-racemic)-Methyl-1-benzyl-4-(2-((tert-butyldimethylsilyloxy)methyl)furo[3,2-b]pyridin-6-yl)pyridine

Supplier: Matrix Scientific

048455-250MG
101883-992EA 393.3 USD
101883-992
(trans-racemic)-Methyl-1-benzyl-4-(2-((tert-butyldimethylsilyloxy)methyl)furo[3,2-b]pyridin-6-yl)pyridine
(trans-racemic)-Methyl-1-benzyl-4-(2-((tert-butyldimethylsilyloxy)methyl)furo[3,2-b]pyridin-6-yl)pyridine

Hazard: Irritant

Formula: C₂₇H₃₆N₂O₄Si
MDL Number: MFCD12922806
CAS Number: 1189172-05-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR