6-Iodo-2-(trimethylsilyl)furo[3,2-b]pyridine

Supplier: Matrix Scientific

048444-250MG
101883-970EA 393.3 USD
101883-970
6-Iodo-2-(trimethylsilyl)furo[3,2-b]pyridine
6-Iodo-2-(trimethylsilyl)furo[3,2-b]pyridine

Hazard: Irritant

Formula: C₁₀H₁₂INOSi
MDL Number: MFCD12922795
CAS Number: 1186310-73-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR