6-Bromo-2-iodofuro[3,2-b]pyridine

Supplier: Matrix Scientific

048440-250MG
101883-962EA 393.3 USD
101883-962
6-Bromo-2-iodofuro[3,2-b]pyridine
6-Bromo-2-iodofuro[3,2-b]pyridine

Hazard: Irritant

Formula: C₇H₃BrINO
MDL Number: MFCD12922791
CAS Number: 1171920-55-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR