2-((tert-Butyldimethylsilyloxy)methyl)furo[3,2-b]pyridin-6-ol

Supplier: Matrix Scientific

048436-250MG
101883-954EA 393.3 USD
101883-954
2-((tert-Butyldimethylsilyloxy)methyl)furo[3,2-b]pyridin-6-ol
2-((tert-Butyldimethylsilyloxy)methyl)furo[3,2-b]pyridin-6-ol

Hazard: Irritant

Formula: C₁₄H₂₁NO₃Si
MDL Number: MFCD12401664
CAS Number: 1171920-47-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR