2-((tert-Butyldimethylsilyloxy)methyl)-6-(trimethylsilyl)furo[3,2-b]pyridine

Supplier: Matrix Scientific

048432-250MG
101883-946EA 393.3 USD
101883-946
2-((tert-Butyldimethylsilyloxy)methyl)-6-(trimethylsilyl)furo[3,2-b]pyridine
2-((tert-Butyldimethylsilyloxy)methyl)-6-(trimethylsilyl)furo[3,2-b]pyridine

Hazard: Irritant

Formula: C₁₇H₂₉NO₂Si₂
MDL Number: MFCD12922785
CAS Number: 1171920-41-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR